3,5-Dichloro-4-fluorobenzoic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC2453-10G
PC2453-1G
PC2453-5G
PC2453-25G
APOSPC2453-10GEA
3640
CZK
APOSPC2453-10G
APOSPC2453-1G
APOSPC2453-5G
APOSPC2453-25G
3,5-Dichloro-4-fluorobenzoic acid 98%
3,5-Dichloro-4-fluorobenzoic acid
Vzorec:
C₇H₃Cl₂FO₂ MW: 209,0019 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03840463 CAS číslo: 98191-30-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...