Tisk…

4'-Bromo-2'-fluoroacetophenone 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-(4-Bromo-2-fluorophenyl)ethanone

PC2391-1G PC2391-25G PC2391-5G
APOSPC2391-1GEA 622 CZK
APOSPC2391-1G APOSPC2391-25G APOSPC2391-5G
4'-Bromo-2'-fluoroacetophenone 98%
4'-Bromo-2'-fluoroacetophenone
Vzorec: C₈H₆BrFO
MW: 217,04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03411551
CAS číslo: 625446-22-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR