4-Bromo-3-fluorobenzoic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC2354-5G
PC2354-100G
PC2354-25G
APOSPC2354-5GEA
622
CZK
APOSPC2354-5G
APOSPC2354-100G
APOSPC2354-25G
4-Bromo-3-fluorobenzoic acid 98%
4-Bromo-3-fluorobenzoic acid
Vzorec:
C₇H₄BrFO₂ MW: 219,01 g/mol Bod tání: 209…212 °C Storage Temperature: Ambient |
MDL Number:
MFCD00672932 CAS číslo: 153556-42-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...