2-(Trifluoromethoxy)phenylacetonitrile 96%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
1-(Cyanmethyl)-2-(trifluormethoxy)benzene, 2-(Trifluoromethoxy)benzyl cyanide
PC2346-1G
PC2346-5G
PC2346-25G
APOSPC2346-1GEA
622
CZK
APOSPC2346-1G
APOSPC2346-5G
APOSPC2346-25G
2-(Trifluoromethoxy)phenylacetonitrile 96%
2-(Trifluoromethoxy)phenylacetonitrile
Vzorec:
C₉H₆F₃NO MW: 201.15 Bod varu: 222…223 °C Hustota: 1.28 g/cm³ (25 °C) Bod vzplanutí: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236326 CAS číslo: 137218-25-8 UN: 3276 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...