Tisk…

1,1,1,2,2,3,3-Heptafluoro-4,6-heptanedione 97%

Dodavatel: Apollo Scientific

PC2345-10G PC2345-5G
APOSPC2345-10GEA 6220 CZK
APOSPC2345-10G APOSPC2345-5G
1,1,1,2,2,3,3-Heptafluoro-4,6-heptanedione 97%
1,1,1,2,2,3,3-Heptafluoro-4,6-heptanedione
Vzorec: C₇H₅F₇O₂
Bod varu: 56…57 °C/38mmHg
Storage Temperature: Ambient
MDL Number: MFCD00511282
CAS číslo: 356-30-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR