Tisk…

3-Bromo-3-chloro-1,1,1-trifluoropropane

Dodavatel: Apollo Scientific

PC2342-5G PC2342-1G
APOSPC2342-5GEA 8570 CZK
APOSPC2342-5G APOSPC2342-1G
3-Bromo-3-chloro-1,1,1-trifluoropropane
3-Bromo-3-chloro-1,1,1-trifluoropropane
Vzorec: C₃H₃BrClF₃
Storage Temperature: Ambient
CAS číslo: 460-66-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR