5-Fluoroindoline 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC2339-1G
PC2339-5G
APOSPC2339-1GEA
1780
CZK
APOSPC2339-1G
APOSPC2339-5G
5-Fluoroindoline 97%
5-Fluoroindoline
Vzorec:
C₈H₈FN MW: 137,16 g/mol Bod varu: 100…102 °C/9mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00214461 CAS číslo: 2343-22-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...