Tisk…

Dibutylammonium tetrafluoroborate 98%

Dodavatel: Apollo Scientific

PC2332-100G PC2332-25G
APOSPC2332-100GEA 11810 CZK
APOSPC2332-100G APOSPC2332-25G
Dibutylammonium tetrafluoroborate 98%
Dibutylammonium tetrafluoroborate
Vzorec: C₈H₂₀BF₄N
Bod tání: 266 °C
Storage Temperature: Ambient
MDL Number: MFCD00034899
CAS číslo: 12107-76-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR