2-Bromo-4-fluorobenzenesulphonyl chloride 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC2305-10G
PC2305-25G
PC2305-5G
APOSPC2305-10GEA
3510
CZK
APOSPC2305-10G
APOSPC2305-25G
APOSPC2305-5G
2-Bromo-4-fluorobenzenesulphonyl chloride 97%
2-Bromo-4-fluorobenzenesulphonyl chloride
Vzorec:
C₆H₃BrClFO₂S MW: 273,51 g/mol Bod varu: 264…265 °C Hustota: 1,87 g/cm³ (25 °C) Bod vzplanutí: 35 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094149 CAS číslo: 351003-45-7 UN: 2921 ADR: 8+4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...