3,5-Bis(trifluoromethyl)toluene 97%
Dodavatel: Apollo Scientific
Synonyms:
1-Methyl-3,5-bis(trifluoromethyl)benzene
PC2296-1G
PC2296-25G
PC2296-5G
APOSPC2296-1GEA
3940
CZK
APOSPC2296-1G
APOSPC2296-25G
APOSPC2296-5G
3,5-Bis(trifluoromethyl)toluene 97%
3,5-Bis(trifluoromethyl)toluene
Vzorec:
C₉H₆F₆ Storage Temperature: Ambient |
MDL Number:
MFCD04972861 CAS číslo: 75462-61-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...