Tisk…

2-Fluoro-1,3-bis-(trifluoromethyl)benzene 97%

Dodavatel: Apollo Scientific

PC2281-25G PC2281-50G PC2281-5G PC2281-1G
APOSPC2281-25GEA 18620 CZK
APOSPC2281-25G APOSPC2281-50G APOSPC2281-5G APOSPC2281-1G
2-Fluoro-1,3-bis-(trifluoromethyl)benzene 97%
2-Fluoro-1,3-bis-(trifluoromethyl)benzene
Vzorec: C₈H₃F₇
Storage Temperature: Ambient
MDL Number: MFCD04972866
CAS číslo: 887268-09-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR