3,5-Dibromo-1-fluorobenzene 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1,3-Dibromo-5-fluorobenzene
PC2260E-100G
PC2260E-25G
PC2260E-500G
APOSPC2260E-100GEA
1120
CZK
APOSPC2260E-100G
APOSPC2260E-25G
APOSPC2260E-500G
3,5-Dibromo-1-fluorobenzene 97%
3,5-Dibromo-1-fluorobenzene
Can be lithiated in 4-position see: Tetrahedron Lett., 37, 6551, 1996
Vzorec:
C₆H₃Br₂F MW: 253,9 g/mol Bod varu: 204…206 °C/768mmHg Hustota: 2,018 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00061119 CAS číslo: 1435-51-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...