Tisk…

1,4-Dibromo-2,5-difluorobenzene 98%

Dodavatel: Apollo Scientific
Varování

PC2235-100G PC2235-25G PC2235-500G
APOSPC2235-100GEA 1580 CZK
APOSPC2235-100G APOSPC2235-25G APOSPC2235-500G
1,4-Dibromo-2,5-difluorobenzene 98%
1,4-Dibromo-2,5-difluorobenzene

see: Org. Synth., 75, 201 (1997), a discussion of lithiation, including dibenzyne formation.

Vzorec: C₆H₂Br₂F₂
Bod varu: 96 °C/20mmHg
Bod tání: 60…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00000346
CAS číslo: 327-51-5
EINECS: 206-317-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR