Tisk…

Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Bis(cycloocta-1,5-dien)rhodium(I)-tetrafluoroborat

PC2198-1G PC2198-250MG
APOSPC2198-1GEA 11580 CZK
APOSPC2198-1G APOSPC2198-250MG
Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate
Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate
Vzorec: C₁₆H₂₄BF₄Rh
MW: 406,072 g/mol
Bod tání: 190 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00075045
CAS číslo: 35138-22-8
EINECS: 460-220-1
UN: 1325
ADR: 4.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR