Tisk…

4-Fluoro-o-phenylenediamine 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3,4-Diamino-1-fluorobenzene, 4-Fluorobenzene-1,2-diamine, 1,2-Diamino-4-fluorobenzene, 4-Fluoro-1,2-phenylenediamine

PC2187-100G PC2187-25G
APOSPC2187-100GEA 4830 CZK
APOSPC2187-100G APOSPC2187-25G
4-Fluoro-o-phenylenediamine 97%
4-Fluoro-o-phenylenediamine

Intermediate for the synthesis of poly(arylene ether phenylquinoxanlines): ACS Symp. Ser. 407, 212, 1999

Vzorec: C₆H₇FN₂
MW: 126.13
Bod varu: 250
Bod tání: 94…98 °C
Bod vzplanutí: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00042228
CAS číslo: 367-31-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR