Tisk…

1,2-Diamino-4-chloro-5-fluorobenzene 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-Chloro-5-fluorobenzene-1,2-diamine

PC2182-1G PC2182-25G PC2182-5G
APOSPC2182-1GEA 622 CZK
APOSPC2182-1G APOSPC2182-25G APOSPC2182-5G
1,2-Diamino-4-chloro-5-fluorobenzene 98%
1,2-Diamino-4-chloro-5-fluorobenzene
Vzorec: C₆H₆ClFN₂
MW: 160,58 g/mol
Bod tání: 110…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00042485
CAS číslo: 139512-70-2
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR