α,α,α-Trifluoro-3,5-diaminotoluene 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
5-(Trifluoromethyl)-1,3-phenylenediamine, 5-(Trifluoromethyl)-m-phenylenediamine, 5-Trifluoromethyl-1,3-phenylenediamine, 5-(Trifluoromethyl)benzene-1,3-diamine, 3,5-Diaminobenzotrifluoride
PC2170-25G
PC2170-5G
PC2170-1G
PC2170-10G
APOSPC2170-25GEA
3580
CZK
APOSPC2170-25G
APOSPC2170-5G
APOSPC2170-1G
APOSPC2170-10G
α,α,α-Trifluoro-3,5-diaminotoluene 98%
α,α,α-Trifluoro-3,5-diaminotoluene
Vzorec:
C₇H₇F₃N₂ MW: 176,14 g/mol Bod tání: 87…89 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014776 CAS číslo: 368-53-6 EINECS: 206-708-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...