2-Fluoro-5-iodobenzoyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC2165-1G
PC2165-25G
PC2165-5G
APOSPC2165-1GEA
1580
CZK
APOSPC2165-1G
APOSPC2165-25G
APOSPC2165-5G
2-Fluoro-5-iodobenzoyl chloride
2-Fluoro-5-iodobenzoyl chloride
Vzorec:
C₇H₃ClFIO MW: 284,46 g/mol Bod tání: 27…30 °C Bod vzplanutí: 59 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03094145 CAS číslo: 186584-73-6 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...