1-[3-(Difluoromethoxy)phenyl]ethanone 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC2132-10G
PC2132-5G
PC2132-25G
APOSPC2132-10GEA
4200
CZK
APOSPC2132-10G
APOSPC2132-5G
APOSPC2132-25G
1-[3-(Difluoromethoxy)phenyl]ethanone 98%
1-[3-(Difluoromethoxy)phenyl]ethanone
Vzorec:
C₉H₈F₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00236220 CAS číslo: 101975-23-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...