Tisk…

α-Bromo-4-chloro-2-fluorotoluene 99%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-Chloro-2-fluorobenzyl bromide, 1-(Bromomethyl)-4-chloro-2-fluorobenzene

PC2119-25G PC2119-5G PC2119-100G PC2119-500G
APOSPC2119-25GEA 992 CZK
APOSPC2119-25G APOSPC2119-5G APOSPC2119-100G APOSPC2119-500G
α-Bromo-4-chloro-2-fluorotoluene 99%
α-Bromo-4-chloro-2-fluorotoluene
Vzorec: C₇H₅BrClF
MW: 223,47 g/mol
Bod varu: 114…118 °C/15mmHg
Hustota: 1,65 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00831105
CAS číslo: 71916-82-0
UN: 3265
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR