Tisk…

Ethyl-5-fluoropyridine-2-carboxylate

Dodavatel: Apollo Scientific

PC210080-1G PC210080-500MG
APOSPC210080-1GEA 12300 CZK
APOSPC210080-1G APOSPC210080-500MG
Ethyl-5-fluoropyridine-2-carboxylate
Ethyl-5-fluoropyridine-2-carboxylate
Vzorec: C₈H₈FNO₂
MW: 169,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11616895
CAS číslo: 148541-70-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR