Tisk…

4-Fluoro-2-iodo-1-nitrobenzene 96%

Dodavatel: Apollo Scientific
Varování

PC210032-25G PC210032-5G
APOSPC210032-25GEA 17630 CZK
APOSPC210032-25G APOSPC210032-5G
4-Fluoro-2-iodo-1-nitrobenzene 96%
4-Fluoro-2-iodo-1-nitrobenzene
Vzorec: C₆H₃FINO₂
Storage Temperature: Ambient
MDL Number: MFCD16658618
CAS číslo: 41860-64-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR