Tisk…

5-Amino-2-(trifluoromethoxy)benzotrifluoride 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-(Trifluoromethoxy)-3-(trifluoromethyl)aniline

PC2034-1G PC2034-250MG PC2034-5G PC2034-100MG
APOSPC2034-1GEA 5590 CZK
APOSPC2034-1G APOSPC2034-250MG APOSPC2034-5G APOSPC2034-100MG
5-Amino-2-(trifluoromethoxy)benzotrifluoride 98%
5-Amino-2-(trifluoromethoxy)benzotrifluoride
Vzorec: C₈H₅F₆NO
MW: 245,12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04972664
CAS číslo: 104678-68-4
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR