Tisk…

2-(4-Chlorophenyl)-3-(trifluoromethyl)pyrazole-4-carbonyl chloride 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC2023-1G PC2023-250MG PC2023-5G
APOSPC2023-1GEA 3540 CZK
APOSPC2023-1G APOSPC2023-250MG APOSPC2023-5G
2-(4-Chlorophenyl)-3-(trifluoromethyl)pyrazole-4-carbonyl chloride 97%
2-(4-Chlorophenyl)-3-(trifluoromethyl)pyrazole-4-carbonyl chloride
Vzorec: C₁₁H₅Cl₂F₃N₂O
MW: 309,08 g/mol
Bod varu: 120 °C/0,3mmHg
Bod tání: 52…53 °C
Storage Temperature: Ambient
MDL Number: MFCD00068143
CAS číslo: 175137-19-6
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR