5-(4-Fluorophenyl)picolinonitrile 98%
Dodavatel: Apollo Scientific
PC201374-250MG
PC201374-1G
PC201374-5G
APOSPC201374-250MGEA
2030
CZK
APOSPC201374-250MG
APOSPC201374-1G
APOSPC201374-5G
5-(4-Fluorophenyl)picolinonitrile 98%
5-(4-Fluorophenyl)picolinonitrile
Vzorec:
C₁₂H₇FN₂ MW: 198,2000 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07782038 CAS číslo: 914349-75-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...