Tisk…

2-(2,4-Dichloro-5-fluorophenyl)piperazine 95+%

Dodavatel: Apollo Scientific

PC201373-10G PC201373-1G PC201373-5G
APOSPC201373-10GEA 18950 CZK
APOSPC201373-10G APOSPC201373-1G APOSPC201373-5G
2-(2,4-Dichloro-5-fluorophenyl)piperazine 95+%
2-(2,4-Dichloro-5-fluorophenyl)piperazine
Vzorec: C₁₀H₁₁Cl₂FN₂
Storage Temperature: Ambient
CAS číslo: 914348-92-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR