1-(3-Chlorophenyl)-2,2,2-trifluoroethylamine 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC201358-250MG
PC201358-1G
APOSPC201358-250MGEA
11580
CZK
APOSPC201358-250MG
APOSPC201358-1G
1-(3-Chlorophenyl)-2,2,2-trifluoroethylamine 95%
1-(3-Chlorophenyl)-2,2,2-trifluoroethylamine
Vzorec:
C₈H₇ClF₃N MW: 209,600 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07374613 CAS číslo: 886368-66-7 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...