Tisk…

2-Fluoro-5-(hydroxymethyl)-6-picoline 95+%

Dodavatel: Apollo Scientific

Synonyms: (6-Fluoro-2-methylpyridin-3-yl)methanol, 6-Fluoro-3-hydroxymethyl-2-methylpyridine

PC201352-100MG PC201352-250MG PC201352-1G
APOSPC201352-100MGEA 2320 CZK
APOSPC201352-100MG APOSPC201352-250MG APOSPC201352-1G
2-Fluoro-5-(hydroxymethyl)-6-picoline 95+%
2-Fluoro-5-(hydroxymethyl)-6-picoline
Vzorec: C₇H₈FNO
MW: 141.1400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05662401
CAS číslo: 884494-98-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR