1-(3-Fluoropyridin-4-yl)ethanone 98%
Dodavatel: Apollo Scientific
PC201340-250MG
PC201340-1G
PC201340-5G
APOSPC201340-250MGEA
3150
CZK
APOSPC201340-250MG
APOSPC201340-1G
APOSPC201340-5G
1-(3-Fluoropyridin-4-yl)ethanone 98%
1-(3-Fluoropyridin-4-yl)ethanone
Vzorec:
C₇H₆FNO MW: 139,1300 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07375077 CAS číslo: 87674-21-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...