6-Fluoro-1H-indazol-3(2H)-one 95+%
Dodavatel: Apollo Scientific
PC201329-250MG
PC201329-1G
APOSPC201329-250MGEA
3150
CZK
APOSPC201329-250MG
APOSPC201329-1G
6-Fluoro-1H-indazol-3(2H)-one 95+%
6-Fluoro-1H-indazol-3(2H)-one
Vzorec:
C₇H₅FN₂O MW: 152,1300 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16038756 CAS číslo: 862274-39-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...