Tisk…

6-Chloro-5-fluoropicolinic acid 97%

Dodavatel: Apollo Scientific

Synonyms: 2-Chloro-3-fluoropyridine-6-carboxylic acid

PC201328-10G PC201328-25G PC201328-5G
APOSPC201328-10GEA 11080 CZK
APOSPC201328-10G APOSPC201328-25G APOSPC201328-5G
6-Chloro-5-fluoropicolinic acid 97%
6-Chloro-5-fluoropicolinic acid
Vzorec: C₆H₃ClFNO₂
Storage Temperature: Ambient
MDL Number: MFCD13185819
CAS číslo: 860296-24-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR