2-Amino-5-bromo-6-fluorobenzonitrile 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
6-Amino-3-bromo-2-fluorobenzonitrile
PC201321-5G
PC201321-1G
PC201321-25G
APOSPC201321-5GEA
1240
CZK
APOSPC201321-5G
APOSPC201321-1G
APOSPC201321-25G
2-Amino-5-bromo-6-fluorobenzonitrile 95%
2-Amino-5-bromo-6-fluorobenzonitrile
Vzorec:
C₇H₄BrFN₂ MW: 215,0225 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06657979 CAS číslo: 845866-92-4 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...