Tisk…

2-Fluoro-m-anisidine 99%

Dodavatel: Apollo Scientific

Synonyms: 2-Fluoro-3-methoxyaniline

PC201312-10G PC201312-1G PC201312-5G
APOSPC201312-10GEA 17160 CZK
APOSPC201312-10G APOSPC201312-1G APOSPC201312-5G
2-Fluoro-m-anisidine 99%
2-Fluoro-m-anisidine
Vzorec: C₇H₈FNO
Bod varu: 231 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11042216
CAS číslo: 801282-00-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR