Tisk…

1-(Piperidin-4-yl)-2-(trifluoromethyl)-1H-benzo[d]imidazole 95+%

Dodavatel: Apollo Scientific

PC201300-250MG PC201300-1G PC201300-5G
APOSPC201300-250MGEA 3610 CZK
APOSPC201300-250MG APOSPC201300-1G APOSPC201300-5G
1-(Piperidin-4-yl)-2-(trifluoromethyl)-1H-benzo[d]imidazole 95+%
1-(Piperidin-4-yl)-2-(trifluoromethyl)-1H-benzo[d]imidazole
Vzorec: C₁₃H₁₄F₃N₃
MW: 269,27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09999148
CAS číslo: 742076-06-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR