Tisk…

3-Fluoro-2,2-dimethylpropanoic acid 97%

Dodavatel: Apollo Scientific

PC201274-100MG PC201274-250MG PC201274-1G
APOSPC201274-100MGEA 1820 CZK
APOSPC201274-100MG APOSPC201274-250MG APOSPC201274-1G
3-Fluoro-2,2-dimethylpropanoic acid 97%
3-Fluoro-2,2-dimethylpropanoic acid
Vzorec: C5H9FO2
MW: 120,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17677039
CAS číslo: 64241-77-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR