Tisk…

4-Fluoro-7-azaindole 97%

Dodavatel: Apollo Scientific

PC201273-1G PC201273-5G
APOSPC201273-1GEA 4860 CZK
APOSPC201273-1G APOSPC201273-5G
4-Fluoro-7-azaindole 97%
4-Fluoro-7-azaindole
Vzorec: C₇H₅FN₂
MW: 136,1300 g/mol
Bod tání: 118…120 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08272231
CAS číslo: 640735-23-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR