Tisk…

2,6-Dichloro-3-fluoropyridine 98%

Dodavatel: Apollo Scientific
Varování

PC201260-5G PC201260-1G
APOSPC201260-5GEA 5590 CZK
APOSPC201260-5G APOSPC201260-1G
2,6-Dichloro-3-fluoropyridine 98%
2,6-Dichloro-3-fluoropyridine
Vzorec: C₅H₂Cl₂FN
Storage Temperature: Refrigerator
MDL Number: MFCD09907815
CAS číslo: 52208-50-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR