Tisk…

3,4-Difluorosalicylaldehyde 95%

Dodavatel: Apollo Scientific

Synonyms: 3,4-Difluoro-2-hydroxybenzaldehyde

PC201259-250MG PC201259-1G
APOSPC201259-250MGEA 870 CZK
APOSPC201259-250MG APOSPC201259-1G
3,4-Difluorosalicylaldehyde 95%
3,4-Difluorosalicylaldehyde
Vzorec: C₇H₄F₂O₂
MW: 158,1023 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08275283
CAS číslo: 502762-95-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR