Tisk…

Methyl-2-fluoroisonicotinate 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Methyl 2-fluoroisonicotinate

PC201256-1G PC201256-5G
APOSPC201256-1GEA 1370 CZK
APOSPC201256-1G APOSPC201256-5G
Methyl-2-fluoroisonicotinate 95%
Methyl-2-fluoroisonicotinate
Vzorec: C₇H₆FNO₂
MW: 155,1300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11656063
CAS číslo: 455-69-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR