4-Fluoro-2-methyl-1H-indol-5-amine 98%
Dodavatel: Apollo Scientific
PC201247-250MG
PC201247-1G
APOSPC201247-250MGEA
1210
CZK
APOSPC201247-250MG
APOSPC201247-1G
4-Fluoro-2-methyl-1H-indol-5-amine 98%
4-Fluoro-2-methyl-1H-indol-5-amine
Vzorec:
C₉H₉FN₂ Storage Temperature: Ambient |
MDL Number:
MFCD09836135 CAS číslo: 398487-76-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...