Tisk…

Methyl-2-(3-amino-2,6-difluorophenyl)acetate 97%

Dodavatel: Apollo Scientific

PC201238-10G PC201238-1G PC201238-5G
APOSPC201238-10GEA 13060 CZK
APOSPC201238-10G APOSPC201238-1G APOSPC201238-5G
Methyl-2-(3-amino-2,6-difluorophenyl)acetate 97%
Methyl-2-(3-amino-2,6-difluorophenyl)acetate
Vzorec: C₉H₉F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD06658469
CAS číslo: 361336-80-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR