Tisk…

2-Fluoro-1-iodo-4-nitrobenzene 95+%

Dodavatel: Apollo Scientific
Varování

PC201225-10G PC201225-5G PC201225-1G
APOSPC201225-10GEA 4960 CZK
APOSPC201225-10G APOSPC201225-5G APOSPC201225-1G
2-Fluoro-1-iodo-4-nitrobenzene 95+%
2-Fluoro-1-iodo-4-nitrobenzene
Vzorec: C₆H₃FINO₂
MW: 267 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00033987
CAS číslo: 2996-30-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR