3-(4-Fluorophenoxy)pyrrolidine hydrochloride 95+%
Dodavatel: Apollo Scientific
PC201207-1G
PC201207-250MG
APOSPC201207-1GEA
4100
CZK
APOSPC201207-1G
APOSPC201207-250MG
3-(4-Fluorophenoxy)pyrrolidine hydrochloride 95+%
3-(4-Fluorophenoxy)pyrrolidine hydrochloride
Vzorec:
C₁₀H₁₃ClFNO Storage Temperature: Ambient |
MDL Number:
MFCD09879244 CAS číslo: 23123-11-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...