3,5-Difluoro-4-(hydroxymethyl)benzonitrile 97+%
Dodavatel: Apollo Scientific
|
Varování
|
PC201204-1G
PC201204-250MG
PC201204-5G
APOSPC201204-1GEA
5100
CZK
APOSPC201204-1G
APOSPC201204-250MG
APOSPC201204-5G
3,5-Difluoro-4-(hydroxymethyl)benzonitrile 97+%
3,5-Difluoro-4-(hydroxymethyl)benzonitrile
Vzorec:
C8H5F2NO MW: 169,1300 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11847159 CAS číslo: 228421-83-8 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...