4-Iodo-2-(trifluoromethyl)aniline 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC2012-1G
PC2012-25G
PC2012-5G
APOSPC2012-1GEA
622
CZK
APOSPC2012-1G
APOSPC2012-25G
APOSPC2012-5G
4-Iodo-2-(trifluoromethyl)aniline 98%
4-Iodo-2-(trifluoromethyl)aniline
Vzorec:
C₇H₅F₃IN Storage Temperature: Ambient |
MDL Number:
MFCD04972657 CAS číslo: 97760-97-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...