7-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline hydrochloride 97%
Dodavatel: Apollo Scientific
PC201199-1G
PC201199-250MG
PC201199-5G
APOSPC201199-1GEA
5890
CZK
APOSPC201199-1G
APOSPC201199-250MG
APOSPC201199-5G
7-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline hydrochloride 97%
7-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline hydrochloride
Vzorec:
C₁₀H₁₁ClF₃N MW: 237,6500 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05861544 CAS číslo: 220247-87-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...