Tisk…

6-Fluorochroman-4-amine hydrochloride 95+%

Dodavatel: Apollo Scientific

PC201188-1G PC201188-250MG
APOSPC201188-1GEA 3310 CZK
APOSPC201188-1G APOSPC201188-250MG
6-Fluorochroman-4-amine hydrochloride 95+%
6-Fluorochroman-4-amine hydrochloride
Vzorec: C₉H₁₁ClFNO
MW: 203,6411 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09026793
CAS číslo: 191609-45-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR