Tisk…
4-[3-(Trifluoromethyl)phenyl]piperidin-4-ol hydrochloride
Dodavatel: Apollo Scientific
Synonyms: 4-(3-(Trifluoromethyl)phenyl)piperidin-4-ol hydrochloride
PC201166-1G PC201166-250MG PC201166-500MG
APOSPC201166-1GEA 8930 CZK
APOSPC201166-1G APOSPC201166-250MG APOSPC201166-500MG
4-[3-(Trifluoromethyl)phenyl]piperidin-4-ol hydrochloride
4-[3-(Trifluoromethyl)phenyl]piperidin-4-ol hydrochloride
Vzorec: C₁₂H₁₅ClF₃NO
MW: 281.71 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00044735
CAS číslo: 1683-49-4