Tisk…

4-(2,2,2-Trifluoroethyl)aniline 95+%

Dodavatel: Apollo Scientific

PC201116-100MG PC201116-1G PC201116-250MG
APOSPC201116-100MGEA 4400 CZK
APOSPC201116-100MG APOSPC201116-1G APOSPC201116-250MG
4-(2,2,2-Trifluoroethyl)aniline 95+%
4-(2,2,2-Trifluoroethyl)aniline
Vzorec: C₈H₈F₃N
Storage Temperature: Ambient
MDL Number: MFCD04972985
CAS číslo: 131395-17-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR