Tisk…

Methyl-4-fluoro-1H-indole-5-carboxylate 95%

Dodavatel: Apollo Scientific

Synonyms: Methyl 4-fluoro-1H-indole-5-carboxylate

PC201093-100MG PC201093-1G PC201093-250MG
APOSPC201093-100MGEA 5690 CZK
APOSPC201093-100MG APOSPC201093-1G APOSPC201093-250MG
Methyl-4-fluoro-1H-indole-5-carboxylate 95%
Methyl-4-fluoro-1H-indole-5-carboxylate
Vzorec: C₁₀H₈FNO₂
MW: 193,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21336690
CAS číslo: 1252782-43-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR